Isobutanol, EMPLURA®, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol

MQ200

1.00985.2500 1.00985.6025 1.00985.9190
1.00985.2500EA 153 GBP
1.00985.2500 1.00985.6025 1.00985.9190
Isobutanol, EMPLURA®, Supelco®
Isobutanol
Formula: (CH₃)₂CHCH₂OH
MW: 74.12 g/mol
Boiling Pt: 107.9
Melting Pt: –108 °C
Density: 0.802 g/cm³ (20 °C)
Flash Pt: 28
Storage Temperature: Ambient
MDL Number: MFCD00004740
CAS Number: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3 III
Merck Index: 13,05148
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de7c83a107c7a71fa54ed2b1bbdfb78617fe3f67&vid=df3ce90fc7ae8dafb7fabe31b244825735325030&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR