Isobutanol, EMSURE® ACS, Reag. Ph. Eur. analysis_for, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol

MQ300

1.00984.1000 1.00984.2500
1.00984.1000EA 122 GBP
1.00984.1000 1.00984.2500
Isobutanol, EMSURE® ACS, Reag. Ph. Eur. analysis_for, Supelco®
Isobutanol
Formula: (CH₃)₂CHCH₂OH
MW: 74.12 g/mol
Boiling Pt: 107.9
Melting Pt: –108 °C
Density: 0.802 g/cm³ (20 °C)
Flash Pt: 28
Storage Temperature: Ambient
MDL Number: MFCD00004740
CAS Number: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3 III
Merck Index: 13,05148
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de7c83a107c7a71fa54ed2b1bbdfb78617fe3f67&vid=df3ce90fc7ae8dafb7fabe31b244825735325030&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 85 g/l (20 °C)
Melting Point -108 °C
Molar Mass 74.12 g/mol
Boiling Point 106 - 108 °C (1013 hPa)
Vapor Pressure 12 hPa (20 °C)
Flash Point 28 °C
Explosion Limit 1.5 - 12 %(V)
Density 0.802 g/cm3 (20 °C)
pH 7 (80 g/l, H2O, 20 °C)
Ignition Point 430 °C
Evaporation number 24


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR