Readybag® Granulated Media for Salmonella and Listeria Control
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.02449.0060 1.00908.0001 1.01865.0001
1.02449.0060EA 581 GBP
1.02449.0060 1.00908.0001 1.01865.0001
Readybag® Granulated Media for Salmonella and Listeria Control
Media for Microbiology Prepared Media for Microbiology
Readybag® gamma-irradiated and pre-weighed granulated media pouches for fast, flexible and easy pathogen testing. Readybag® media pouches speed up and simplify food pathogen testing routines: All media preparation steps are eliminated from workflow. Just add sterile water before incubation. Readybag® series includes Salmonella (Buffered Peptone Water) and Listeria (Half Fraser Broth).
  • No weighing
  • No autoclaving
  • No supplement handling
  • No need for extra lab space or equipment