Ethyl acetate, Uvasol® for spectroscopy, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Acetic acid ethyl ester

M-Clarity™ quality level = MQ100

1.00863.0500 1.00863.2500
1.00863.0500EA 43.33 GBP
1.00863.0500 1.00863.2500
Ethyl acetate, Uvasol® for spectroscopy, Supelco®
Ethyl acetate
Formula: CH₃COOC₂H₅
Boiling Pt: 77.1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 85.3 g/L (20 °C)
Melting Point -83 °C
Molar Mass 88.11 g/mol
Boiling Point 77 °C (1013 hPa)
Vapor Pressure 97 hPa (20 °C)
Flash Point -4 °C
Refractive Index 1.37
Explosion Limit 2.1 - 11.5%(V)
Density 0.90 g/cm³ (20 °C)
pH (H₂O) no data available
Ignition Point 460 °C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR