Methenamine silver plating kit acc. Gomori for histology

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

1.00820.0001
1.00820.0001EA 325 GBP
1.00820.0001
Methenamine silver plating kit acc. Gomori for histology
Methenamine silver plating kit

Gomori’s Methenamine Silver stain is used on basal membranes, bacteria and fungi for the detection of argent-affine structures in histological tissue.


The Methenamine silver plating kit acc. to Gomori contains five ready-to-use solutions and methenamine/borate tablets, whereby one tablet is sufficient for the preparation of 30 ml of ready-to-use silver solution. A microwave protocol has been compiled for laboratories that use a microwave, which is available on request.The package is sufficient for up to 50 applications. The kit is an IVD product and CE certified, it can be used in the accredited diagnostic laboratory.


The target structures are turned black, while everything else is stained green with Light green SF solution. The color of the silver deposits can be intensified still further by treatment with gold chloride solution, through formation of a silver-gold complex. The use of gold chloride solution also prevents non-specific staining of the background.

Storage Temperature: Ambient
UN: 3316
ADR: 9 II
IVDLogo

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR