Water, SupraSolv® for headspace gas chromatography, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Hydrogen oxide

M-Clarity™ quality level = MQ100

1.00577.2500 1.00577.1000
1.00577.2500EA 120.75 GBP
1.00577.2500 1.00577.1000
Water, SupraSolv® for headspace gas chromatography, Supelco®
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Molar Mass 18.02 g/mol
Boiling Point 100 °C (1013 hPa)
Vapor Pressure 23 hPa (20 °C)
Density 1.00 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR