TLC plates, silica gel 60G, glass backed
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.00384.0001 1.00390.0001
1.00384.0001EA 324 GBP
1.00384.0001 1.00390.0001
TLC plates, silica gel 60G, glass backed
TLC Plates
Highly robust plates with gypsum as binder, fully compliant with international pharmacopoeia (Ph. Eur. and USP).
Traditionally, TLC monographs in a pharmacopoeia refer to products using silica G, containing gypsum as binder, or silica H with no foreign binder. There are about 200 monograph methods in the European Pharmacopoeia (Ph. Eur.) referring to these plates*. Fully compliant with international pharmacopoeia, these new TLC silica gel 60G plates are recommended for customers in QA/QC labs using older Ph. Eur. monograph methods, which require TLC plates with a gypsum binder and who doMore Product Information