Ammonium iron (III) citrate (14,5 - 16,0% Fe) (RT), green, powder, Purum p.a.

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Ammonium ferric citrate, Ferric Ammonium Citrate, Ammonium iron (III) citrate

09713-1KG 09713-500G 09713-50G
09713-1KGEA 123 GBP
09713-1KG 09713-500G 09713-50G
Ammonium iron (III) citrate (14,5 - 16,0% Fe) (RT), green, powder, Purum p.a.
Ammonium iron (III) citrate

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formula: C₆H₅+4y)Fe(x)N(y)O₇
Storage Temperature: Ambient
MDL Number: MFCD00013099
CAS Number: 1185-57-5
EINECS: 214-686-6
Merck Index: 13,00518

Order Now

Specification Test Results

Assay of Fe 14.5 - 16.0 %
Calcium (Ca) Max. 1000 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Potassium (K) Max. 500 ppm
Sodium (Na) Max. 5000 ppm
Nickel (Ni) Max. 100 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Chloride (Cl) Max. 500 ppm
Sulfate (SO₄) Max. 500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR