Barium carbonate 99.0-101.0% (by acidimetry), powder, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

0950-REA 15580 GBP
0950-R 0950-01 0950-07
Barium carbonate 99.0-101.0% (by acidimetry), powder, BAKER ANALYZED® ACS, J.T.Baker®
Barium carbonate

Powder. Lot analysis on label.

Formula: BaCO₃
Melting Pt: 1350 °C
Density: 4.30 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1,III
Merck Index: 13,00972

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements 99.0 - 101.0%
For Laboratory,Research,or Manufacturing Use ≤0.015%
Assay (BaCO₃) (by acidimetry) 99.0 - 101.0 %
Insoluble in Dilute HCl ≤ 0.015 %
Chloride (Cl) ≤ 0.002 %
Water-Soluble Titrable Base, meq/g ≤ 0.0020
Oxidizing Substances (as NO₃) ≤ 0.005 %
Sulfide (S) ≤ 0.001 %
Calcium (Ca) ≤ 0.05 %
Heavy Metals (as Pb) ≤ 0.001 %
Iron (Fe) ≤ 0.002 %
Potassium (K) ≤ 0.005 %
Sodium (Na) ≤ 0.010 %
Strontium (Sr) ≤ 0.7 %
Average Particle Diameter, µm (APD) ≤ 5
Specific Surface Area, m/g, For Information Only
Bulk Density (g/cc), For Information Only
Mesh (Wet Screen Analysis) - On U.S. No. 325 Sieve, For Information Only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR