Supelco® Organic Reference Standard, 2,4-Dichlorophenol
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
07846-100MG
07846-100MGEA 105 GBP
07846-100MG
Supelco® Organic Reference Standard, 2,4-Dichlorophenol
Standards Organic Standards
  • GHS signal: GHS06, GHS05, GHS09
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P260 - P273 - P280 - P301 + P312 - P303 + P361 + P353 - P305 + P351 + P338
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55936411a0dbf2c2b46a6beaa6b96bafb8c8996c&vid=55936411a0dbf2c2b46a6beaa6b96bafb8c8996c&prid=web&clid=SAPDAM