Ammonium dihydrogen phosphate 98,0, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

0776-01EA 94 GBP
0776-01 0776-07 0776-R
Ammonium dihydrogen phosphate 98,0, BAKER ANALYZED® ACS, J.T.Baker®
Ammonium dihydrogen phosphate
Formula: NH₄H₂PO₄
MW: 115.03 g/mol
Melting Pt: 190
Density: 1.80 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1
EINECS: 231-764-5
Merck Index: 13,00547

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements ≥98.0%
For Laboratory,Research,or Manufacturing Use ≤0.001%
Assay (NH₄H₂PO₄) ≥98.0 %
Calcium (Ca) ≤0.001 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 3.8 - 4.4
Magnesium (Mg) ≤0.0005 %
Nitrate (NO₃) ≤0.001 %
Sulfate (SO₄) ≤0.01 %
Trace Impurities - Iron (Fe) ≤0.001 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Chloride (Cl) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR