Hydrogen chloride solution 0.5 M in methanol, LiChropur™ for GC derivatisation, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

07607-10X1ML 07607-100ML
07607-10X1MLEA 54.8 GBP
07607-10X1ML 07607-100ML
Hydrogen chloride solution 0.5 M in methanol, LiChropur™ for GC derivatisation, Supelco®
Hydrogen chloride

Methanolic HCl was used in fatty acid analysis during a study to identify Δ4 fatty acid desaturated from Thraustochytrium sp. involved in the biosynthesis of docosahexanoic acid.

Formula: HCl
Density: 0.800...0.811 g/m³ (20 °C)
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 3286
ADR: 3 (6.1, 8),II

Order Now

Specification Test Results

Appearance (Color) Colorless
Appearance (Form) Liquid
Titration (Method) Alkalimetric
Titration (Molarity) 0.45 - 0.55 mol/l
Remarks on GC Suitibility for GC derivatization complying
Density D20/4 0.800 - 0.811
Proton NMR Spectrum Conforms to Structure
Recommended Retest Period 54 Months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR