, anhydrous 98 ACS

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

0705-2.5KGEA 405 GBP
0705-2.5KG 0705-500G 0705-12KG 0705-50KG
, anhydrous 98 ACS
di-Potassium hydrogen phosphate
Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chloride <0.003%
Heavy Metals (as Pb) ≤0.0005%
Insolubles ≤0.01%
Iron ≤0.001%
Loss on Drying ≤1%
Nitrogen Compounds ≤0.001%
pH (5%, Water) @25°C 8.5 – 9.6
Purity ≥98%
Sodium ≤0.05%
Sulphate ≤0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR