Aluminium chloride, anhydrous ≥99.0% (by argentometric titration), crystallised, Puriss. p.a., Fluka™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Aluminum chloride

06220-1KG
06220-1KGEA 157 GBP
06220-1KG
Aluminium chloride, anhydrous ≥99.0% (by argentometric titration), crystallised, Puriss. p.a., Fluka™
Aluminium chloride
Formula: AlCl₃
MW: 133.34 g/mol
Boiling Pt: 180
Melting Pt: 190 °C (2.500 hPa)
Density: 2.44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003422
CAS Number: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8 II
Merck Index: 13,00335

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 10 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 10 ppm
Magnesium (Mg) Max. 10 ppm
Sodium (Na) Max. 10 ppm
Nickel (Ni) Max. 10 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 10 ppm
Sulfate (SO₄) Max. 100 ppm
Vapor Pressure 1 mmHg ( 100 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR