Acetylacetone, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: ACAC, 2,4-Pentanedione

MQ100

05581-1ML-F 05581-5ML-F
05581-1ML-FEA 43.2 GBP
05581-1ML-F 05581-5ML-F
Acetylacetone, Supelco®
Acetylacetone
Formula: CH₃COCH₂COCH₃
MW: 100.12 g/mol
Boiling Pt: 140.4
Melting Pt: –23 °C
Density: 0.96945 g/cm³ (20 °C)
Flash Pt: 30
Storage Temperature: Ambient
MDL Number: MFCD00008787
CAS Number: 123-54-6
EINECS: 204-634-0
UN: 2310
ADR: 3 III
Merck Index: 13,00083
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d19f64503f6ec35059189727d6f68d7d94826c35&vid=988d7a5387ef3c322a4d7be99e1d67829f97153a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR