Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.9%, Ultrapure

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM

0497-1KGEA 203 GBP
0497-1KG 0497-5KG 0497-500G 0497-10KG 0497-12KG 0497-50KG
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.9%, Ultrapure
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)

Reagent for stabilisation of buffer formulations and electrophoresis of biological molecules.


Biological buffers are useful for cell culture in vitro, enzyme assays and some electrophoretic applications at physiological pH. Universally applicable buffers for biochemistry must be water soluble, not interfere with biological processes or biological membranes (penetration, solubilisation, adsorption on surface, etc.), have known complex forming tendency with metal ions, be non toxic and have a very low U.V. absorption at wavelength >260 nm.

Formula: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Boiling Pt: 288 °C
Melting Pt: 169 °C
Density: 1.328 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@280 nm (1 M, Water) <0.05
Abs.@290 nm (40%, Water) ≤0.2
Abs.@400 nm (40%, Water) ≤0.05
Arsenic ≤0.0005%
Copper ≤0.0001%
Insolubles ≤0.005%
Iron ≤0.0001%
Lead ≤0.0001%
Magnesium ≤0.0005%
Melting Point 168 – 172 °C
Moisture (KF) ≤1%
pH (5%, Water) @25 °C 10 - 11.5
Purity ≥99.9%
Solubility (10%, Water) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR