Florisil® for chromatography, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil

03286-100G-F 03286-500G-F
03286-100G-FEA 107 GBP
03286-100G-F 03286-500G-F
Florisil® for chromatography, Supelco®
Magnesium silicate
Formula: MgO₃Si
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58d552e905e9b14e6fe1cb8ab8103a9b3e788b68&vid=58d552e905e9b14e6fe1cb8ab8103a9b3e788b68&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR