VWR® UltraPure, Tris-glycine buffers, 10X concentrate
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0307-4LEA 260 GBP
0307-4L 0307-1L 0307-5L
VWR® UltraPure, Tris-glycine buffers, 10X concentrate
Buffers Electrophoresis Buffers Running Buffers
Running buffers for both 1-D and 2-D protein gel electrophoresis.
  • Concentrated solutions with a choice of packaging options.
A 1X working solution contains 0,025 M Tris base and 0,192 M glycine.