Bovine native Casein (from Milk), MP Biomedicals
Supplier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0290129301 0290129305 0290129325 0290129380
ICNA0290129301EA 72.7 GBP
ICNA0290129301 ICNA0290129305 ICNA0290129325 0290129380.
Bovine native Casein (from Milk), MP Biomedicals
Proteins and Peptides
Casein from bovine milk is a phosphoprotein. It make up approximately 80% of the total protein in bovine milk. It is proposed to be the main protective constituent in milk.
It has a wide variety of uses, from being a major component of cheese, to use as a food additive, to a binder for safety matches. As a food source, casein supplies amino acids; carbohydrates; and two inorganic elements, calcium and phosphorus.
It is produced by controlled acidification of pure, pasteurized skim milk, resulting in precipitation of casein curd. The curd is then washed, dried, and ground to a consistent particle size.