Pyridine-2,6-dicarboxylic acid, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ100

02321-10G-F 02321-5G-F
02321-10G-FEA 87.7 GBP
02321-10G-F 02321-5G-F
Pyridine-2,6-dicarboxylic acid, Supelco®
Pyridine-2,6-dicarboxylic acid
Formula: C₇H₅NO₄
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 327 °C
Flash Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00006299
CAS Number: 499-83-2
EINECS: 207-894-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f034b0b927063a28f5e4f16b4ce8cc981b4a391&vid=f3565b2c5d354404461b9e68cf369ea9c79b62f8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR