Succinic acid 99, high_purity

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1,4-Butanedioic acid

0165-2.5KGEA 194 GBP
0165-2.5KG 0165-500G
Succinic acid 99, high_purity
Succinic acid

Biological buffers are useful for cell culture in vitro, enzyme assays and some electrophoretic applications at physiological pH.

Formula: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 182 °C
Density: 1.56 g/cm³ (20 °C)
Flash Pt: 206
Storage Temperature: Ambient
MDL Number: MFCD00002789
CAS Number: 110-15-6
EINECS: 203-740-4
UN: 3261
ADR: 8 III
Merck Index: 13,08953
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfd381e6e0e4cbde676a92c9e8d6aacfac635ade&vid=0b14e1b7f82b03468fc82d224a0ee894952eb200&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Heavy Metals (as Pb) ≤0.001%
Melting Point 185 – 190 °C
Purity > 99%
Residue on Ignition ≤0.025%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR