D-(+)-Glucose monohydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0113.1000EA 38 GBP
0113.1000 0113.9050
D-(+)-Glucose monohydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
D(+)-Glucose monohydrate
Formula: C₆H₁₂O₆·H₂O
Melting Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00149450
CAS Number: 14431-43-7
EINECS: 200-075-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9ea2af2ca62ca94d86809070593a60aafd8cd1b&vid=e549b499dca7fb95811640efad1429268c8f6d8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chloride (Cl) max. 0.005 %
Insoluble Matter max. 0.005 %
Residue after Ignition max. 0.015 %
Starch passes test
Sulfate and Sulfite (as SO4) max. 0.005 %
Trace Impurities (in ppm)Arsenic (As) max. 0.2
Trace Impurities (in ppm)Iron (Fe) max. 5
Trace Impurities (in ppm)Heavy Metals (as Pb) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR