Titanium ≥93%, powder <20 µm

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

00681.14 00681.22 00681.36 00681.06
00681.14EA 85.2 GBP
00681.14 00681.22 00681.36 00681.06
Titanium ≥93%, powder <20 µm
Titanium

Purity: 93% CAS No.: 7440-32-6 Physical Form: Packaged in water Odour: Odourless UN No.: 1352 MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3277 °C (1013 hPa)
Melting Pt: 1677 °C
Density: 4.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
UN: 1352
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR