Ruthenium(III) chloride oxide 85.0-115.0% (dried basis, AT) for microscopy, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Ruthenium oxychloride, Tetraamminechlorohydroxoruthenium(III) chloride, Ruthenium Red

00541-1G 00541-5G
00541-1GEA 233 GBP
00541-1G 00541-5G
Ruthenium(III) chloride oxide 85.0-115.0% (dried basis, AT) for microscopy, Sigma-Aldrich®
Ruthenium(III) chloride oxide

(Ammoniated ruthenium oxychloride)

Formula: H₄₂Cl₆N₁₄O₂Ru₃
MDL Number: MFCD00011479
CAS Number: 11103-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR