Titanium ≥99.98% (metals basis), wire, Ø 0.75 mm (0.03 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

00361.BU 00361.BY
00361.BUEA 59.3 GBP
00361.BU 00361.BY
Titanium ≥99.98% (metals basis), wire, Ø 0.75 mm (0.03 in)
Titanium

Purity: 99.98% CAS No.: 7440-32-6 Physical Form: ≈1.99g/m, Temper: as drawn Odour: Odourless MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR