Niobium ≥99.97% (metals basis excluding Ta), foil (max. 0.06% Ta), Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

00238.FF 00238.FQ
00238.FFEA 81.1 GBP
00238.FF 00238.FQ
Niobium ≥99.97% (metals basis excluding Ta), foil (max. 0.06% Ta), Thickness: 0.25 mm (0.01 in)
Niobium

Purity: 99.97% CAS No.: 7440-03-1 Physical Form: ≈1.34g/25x25mm Appearance: Grey or silver Odour: Odourless MDL No.: MFCD00011126

Formula: Nb
Boiling Pt: 3700 °C (1013 hPa)
Melting Pt: 2468 °C
Density: 8.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR