Ammonium chloride, BAKER, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

0019.1000EA 50.4 GBP
0019.1000 0019.9050 0019.9025BB
Ammonium chloride, BAKER, J.T.Baker®
Ammonium chloride
Formula: NH₄Cl
Melting Pt: 335 °C
Density: 1.5256 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets Reagent Specifications for testing Ph. Eur. monographs.
Assay (dry basis) 99.5 - 100.5 %
Acidity or Alkalinity passes test
Appearance of solution passes test
Bromides and Iodides passes test
Calcium (Ca) max. 200 ppm
Iron (Fe) max. 20 ppm
Heavy Metals (as Pb) max. 10 ppm
Identification passes test
Loss on Drying max. 0.5 %
Sulfated Ash max. 0.1 %
Sulfates (as SO4) max. 150 ppm
Additional Specification(s):
Limit of thiocyanate (SCN) passes test
pH of 5% Solution at 25°C 4.6 - 6.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR