Safety footwear
uvex safety footwear with protection class S1, S1 P, S2, S3 and S5 (EN ISO 20345) provides perfect protection in industry sectors such as pharma, biopharma, food and beverage, education, production, chemical industry, electronics and aerospace. Our 'uvex' range also includes occupational footwear (protection class O1, EN ISO 20345) and tailor-made orthopaedic safety footwear.
uvex 1 sport safety shoes - pink, blue or black
ESD lace-up safety shoes. Lightweight, flexible and metal-free. In black and two eye-catching colour options.
uvex 1 safety shoes
Lace-up ESD safety shoes with water resistant high-tech micro velour upper and Climazone technology.
uvex 2 VIBRAM leather safety shoes
Robust ESD lace-up safety shoes with dual-density, slip-resistant VIBRAM® rubber soles.
uvex 2 GTX safety boots
Watertight and highly breathable ESD safety boot with VIBRAM® anti-slip sole.
uvex 2 Xenova
Lightweight all round shoe with breathable upper materials and comfortable insole
uvex 1 x-tended support
Ergonomically designed safety shoe for outstanding wearer comfort and maximum impact protection
Videos
Literature
Twisting, stumbling, slipping
What happens? Why? And what can be done to prevent it?