Dew-point and humidity meters
For certain specific applications in the pharmaceutical industry, the calibration must be carried out every 3 months, as measuring devices are subject to the validation process and even the smallest fluctuations can have a large influence on the application.
Humidity calibrator
- For calibration of humidity sensors, transmitters and portable devices
- Generates stable humidity values of 10 to 90% RH
- Compact and easy to transport
- Rapid stabilisation of the test chamber
- Optional: independent traceable reference hygrometer
- Several sensors can be calibrated simultaneously
Humidity / temperature calibrator
- For calibration of rF humidity sensors and transmitters
- Temperatures from 10 to 50 °C with a precision of 0.1 °C and humidity of 10 to 90% RH
- Precise and repeatable calibrations are possible
- Generates humidity and temperature simultaneously
- Excellent stability
- Compact and easy to transport
- Optional: built-in data recorder, logs the built-in reference sensor and the sensors to be calibrated
- Suitable for sensors with Ø betw. 5 and 25 mm