A major plus for routine analysis
![](/m-uk.vwr.com/en_GB/images/Just_it_image200x200.jpg)
New Spectroquant® Prove plus spectrophotometers with added speed, storage, and simplicity
As your trusted partner in analytical chemistry, we’re always working on the next innovation to enhance your analytical work. Our latest generation of spectrophotometers for your analysis does just that. The Spectroquant® Prove plus range is packed with features that make accuracy and precision simpler than ever.
Positives of Prove plus for water, environmental and food & beverage analysis
- Smoother functionality with an instantly responsive display and faster processing
- Greater capacity to store more measurement results (up to 7000)
- Extended warranty period of two years
Features and Benefits
Pre-programmed to work seamlessly with more than 150 Spectroquant® test kits, Spectroquant® Prove plus photometers are among the most trusted for your analysis.
- Easy to use and pre-programmed with pre-defined reagents
- Reduced use of chemicals compared to traditional methods
- Suitable for a broad measuring range per parameter (minimizes dilution work)
- Pre-programmed for over 300 analysis methods
- Cell size up to 10 cm with Prove 600 plus for high-sensitivity measurements
- Instrument-supported AQA system for easy quality assurance
- Automatic cell size recognition for cell & reagent tests
- Live ID barcode for all cell & reagent tests (incl. lot and expiry data)
- No adapter needed for the measurement of different cell sizes
- Kits can be conveniently stored at room temperature
Spectroquant® PROVE 100 | Spectroquant® PROVE 300 | Spectroquant® PROVE 600 |
![]() |
![]() |
![]() |
Prove 100 plus | Prove 300 plus | Prove 600 plus |
Routine VIS measurements in wastewater analysis and environmental testing Examples: Groundwater, effluents, soil |
Sensitive UV/VIS analysis of drinking water, food and beverages Examples: Drinking water, beverages, special beer applications, sugar, olive oil |
Complex UV/Vis measurements in process water testing Examples: Power plants, cooler & boiler water |
1.73026.0001 | 1.73027.0001 | 1.73028.0001 |
Introduction
How to start the Prove
How to navigate through the functions
How to use Prove with the Spectroquant® cell test kits and reagent sets
How to easily clean the Prove
How to select the language, date and time
Datatransfer - Data on demand - easy to filter - easy to share
How to check interfering matrix
Be sure of your system: Use AQA Analytical Quality Assurance