Transparent Paper Cleanroom Packaging
Features:
- Material: sterilisation kraft paper with composite foil of polyester/polypropylene
- Transparent foil offers the advantage of product visibility
- Unlimited suitability for steam-, ethylene-oxide- and formaldehyde-sterilisation
- Sterilisation indicators on the packaging
- Simple, safe sealing with pass-through sealing devices
- Removal help facilitates opening of the packaging
- Foils are suitable for printing with sterilisable fibre-tipped pens
- Optionally available as flat paper bag or tube (ideal for oversized goods)
Product selection
Transparent Paper Packaging
Please choose from a list of products and refine your search by attributes...