Cleanroom Computer Accessories
Since the keyboard is one of the areas in the cleanroom with the highest level of contamination and is very difficult to clean, many users cover it with a protective foil. The specially developed keyboard for use in cleanrooms is a tremendous improvement over such foils.
The touch-sensitive cleanroom keyboards have no gaps between the keys to reduce collection of particles. The cleanroom keybord is completely sealed and waterproof and therefore easy to clean and to disinfect. Made of acrylic or glass. Suitable for complete infection control.
Cleanroom mouses have exactly the same functionality as an ordinary desktop mouse. The cleanroom mouse is sealed and for thorough disinfecting, it can be completely immersed in a cleaning solution. The mouses are compatible with any USB connection and with all computer systems.