Harvesting Process
Temperature Sensors
Micro-Filtration
Depending on the production floor or laboratory throughput, the filter will range in capacity and act as a rough filter in the microscale.
Ultra-Filtration
One of the final steps in the harvest workflow, ultrafiltration acts to remove the finest impurities from the target macromolecule product resulting a concentrate. This is usually done at a high pressure.