Merck 3S reagents for titration with Metrohm OMNIS
![](/m-uk.vwr.com/en_GB/images/merck_3s_reagents_main_200x200.jpg)
3S is an intelligent packaging solution protected with an originality seal. There is an automatic data transfer between the cap and the Metrohm OMNIS titrator using RFID (radio-frequency identification).
Safe Safe chemical handling: minimized risk of contamination through contact-free reagent exchange
Smart Smart connection of chemical consumables: communication between the 3S cap and instrument software via RFID with a visual status indicator on the system
Secure Secure information transfer: Direct RFID-based transfer of reagent data to instrument software for elimination of human error. How to use
- Remove tamper evident ring and push down to open
- Connect with a single hand movement to the Omnis liquid adapter
- Reagent data is automatically transferred to the method