Quantus Under Garments
In order to optimise the Quantus® garment system, we recommend the use of abrasion-proof, functional under garments (Underwear).
High quality blends in polyester, microfibre or cotton/polyester that minimise loose fibres are available. Additional comfort is achieved through optimal moisture and heat transport.
Besides physiological aspects and comfort, aspects of hard-wearing work clothes as well as CI-requirements and modern styles and colours are becoming more and more important when choosing under garments.
The product range of Quantus under garments includes:
- Sweatshirts (long sleeves)
- T-shirts (short sleeves)
- Trousers
- Jackets
- Socks
Fabrics for Quantus Under Garments
Model Overview Quantus Under Garments
Sizing for Quantus Under Garments
Underwear, standard
Sweatshirt (long sleeves with knitted cuffs), T-shirts (short sleeves), trousers (with rubber band at ankle height) available. Made of microfibre, antimicrobial finish
AreaDress
Top and Trousers. Antistatic and antimicrobial equipped. Keenly softer feeling, with tightly texture tie and excellent breathability.
Socks
Socks with low abrasion and washable to 30 °C.