Permanent Cleanroom Mats
Solvent-based cleaning materials should on no account be used on these mats: they will adversely affect both the function and the appearance. To avoid risk of slipping, it is also important that both the mats and the flooring should be completely dry.
Product selection
Permanent Mats
Please choose from a list of products and refine your search by attributes...