Tyvek Cleanroom Packaging
Features:
- Composite of polyethylene foil and uncoated Tyvek®
- Transparent foil offers the advantage of product visibility
- Steam-sterilisable up to 125 °C
- Ethylene oxide- and gamma sterilisable
- Very low gas residue (for instance ETO) gas sterilisation shortens the airing out time
- Sterilisation indicators on the packaging
- Extremely low particle contamination
- Outstanding barrier characteristics
- High-quality sealing and burst strength
- Easily heat-sealed (sealing temperature up to max. 120 °C)
- Foils are suitable for printing
- 100% recyclable
- Optionally available as flat bag or easy-tear-bag to minimise particle contamination
- Available in various cleanroom classes
Product selection
.png)
Tyvek-Packaging
Please choose from a list of products and refine your search by attributes...