Cleanroom Knives and Bag Cutter
Special knives for easy opening of packaging and other cutting work. Available as reusable or disposable versions.
The reusable version has an automatically retractable safety blade guard. No blade changes required - eliminates all loose blades. With ergonomic handle and gamma irradiated.
The reusable version is made of stainless steel, autoclavable with replaceable blade.
Product selection
Knives and Cutters
Please choose from a list of products and refine your search by attributes...