cis-Jasmone
Supplier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(Z)-3-Methyl-2-(pent-2-en-1-yl)cyclopent-2-enone
, cis-Jasmon
ROTH6519.1EA
0
GBP
ROTH6519.1
cis-Jasmone
cis-Jasmone
Formula:
C₁₁H₁₆O MW: 164.25 g/mol |
MDL Number:
MFCD00001402 CAS Number: 488-10-8 EINECS: 207-668-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...