4-Methoxy-3-(trifluoromethyl)benzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
005829-5G
005829-25G
005829-2.5G
5829-25G
FLUO005829-5GEA
0
GBP
FLUO005829-5G
FLUO005829-25G
FLUO005829-2.5G
FLUO5829-25G
4-Methoxy-3-(trifluoromethyl)benzonitrile
4-Methoxy-3-(trifluoromethyl)benzonitrile
Formula:
C₉H₆F₃NO MW: 201.15 g/mol |
MDL Number:
MFCD01631575 CAS Number: 261951-87-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...