4-Methoxynicotinic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7268-1G
PY-7268-5G
COBBPY-7268-1GEA
0
GBP
COBBPY-7268-1G
COBBPY-7268-5G
4-Methoxynicotinic acid
4-Methoxynicotinic acid
Formula:
C₇H₇NO₃ MW: 153.1400 g/mol |
MDL Number:
MFCD04115724 CAS Number: 10177-31-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...