2,5-Bis(trifluoromethyl)benzaldehyde, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

463930-10G 463930-1G
SIAL463930-10GEA 318 GBP
SIAL463930-10G SIAL463930-1G
2,5-Bis(trifluoromethyl)benzaldehyde, Sigma-Aldrich®
2,5-Bis(trifluoromethyl)benzaldehyde
Formula: C₉H₄F₆O
MW: 242.12 g/mol
MDL Number: MFCD00674087
CAS Number: 395-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR