3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbothioamide
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008241-1G
008241-5G
FLUO008241-1GEA
0
GBP
FLUO008241-1G
FLUO008241-5G
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbothioamide
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbothioamide
Formula:
C₁₁H₈ClFN₂S |
MDL Number:
MFCD00052560 CAS Number: 175204-42-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...