3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008240-1G 008240-5G
FLUO008240-1GEA 0 GBP
FLUO008240-1G FLUO008240-5G
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxamide
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxamide
Formula: C₁₁H₈ClFN₂O₂
MDL Number: MFCD00173954
CAS Number: 4415-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR