D-(-)-Isoascorbic acid
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(R)-5-((R)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one
, Erythorbic acid
, D-Isoascorbic acid
EHERC14379000EA
94.4
GBP
EHERC14379000
D-(-)-Isoascorbic acid
D(-)-Isoascorbic acid
Formula:
C₆H₈O₆ MW: 176.12 g/mol |
MDL Number:
MFCD00005378 CAS Number: 89-65-6 EINECS: 201-928-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...