2-Fluoro-4-methyl-5-pyridineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-4-methylpyridine-5-boronic acid

BB-8568-5G BB-8568-1G
COBBBB-8568-5GEA 0 GBP
COBBBB-8568-5G COBBBB-8568-1G
2-Fluoro-4-methyl-5-pyridineboronic acid
2-Fluoro-4-methyl-5-pyridineboronic acid
Formula: C₆H₇BFNO₂
MW: 154.9400 g/mol
MDL Number: MFCD08741406
CAS Number: 1072944-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR