2-Methyl-4-pyridineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Methylpyridin-4-yl)boronic acid, 2-Methylpyridine-4-boronic acid

BB-5249-1G BB-5249-5G
COBBBB-5249-1GEA 0 GBP
COBBBB-5249-1G COBBBB-5249-5G
2-Methyl-4-pyridineboronic acid
2-Methyl-4-pyridineboronic acid
Formula: C₆H₈BNO₂
MW: 136.9442 g/mol
MDL Number: MFCD03095364
CAS Number: 579476-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR