2-Methoxy-5-pyridineboronic acid hydrochloride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-2089-5G
BB-2089-1G
COBBBB-2089-5GEA
0
GBP
COBBBB-2089-5G
COBBBB-2089-1G
2-Methoxy-5-pyridineboronic acid hydrochloride
2-Methoxy-5-pyridineboronic acid hydrochloride
Formula:
C₆H₉BClNO₃ |
MDL Number:
MFCD04973740 CAS Number: 370864-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...