2-Methoxy-3-pyridineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Methoxypyridine-3-boronic acid

BB-2328-5G BB-2328-25G BB-2328-1G
COBBBB-2328-5GEA 0 GBP
COBBBB-2328-5G COBBBB-2328-25G COBBBB-2328-1G
2-Methoxy-3-pyridineboronic acid
2-Methoxy-3-pyridineboronic acid
Formula: C₆H₈BNO₃
MW: 152.95 g/mol
MDL Number: MFCD03411572
CAS Number: 163105-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR