2-Morpholino-5-pyridineboronic acid pinacol ester, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-(4-Morpholinyl)pyridine-3-boronic acid pinacol ester, 2-(Morpholin-4-yl)pyridine-5-boronic acid pinacol ester, 4-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)morpholine

M-Clarity™ quality level = MQ100

654310-1G 654310-5G
SIAL654310-1GEA 61 GBP
SIAL654310-1G SIAL654310-5G
2-Morpholino-5-pyridineboronic acid pinacol ester, Sigma-Aldrich®
2-Morpholino-5-pyridineboronic acid pinacol ester
Formula: C₁₅H₂₃BN₂O₃
MW: 290.17 g/mol
MDL Number: MFCD04039874
CAS Number: 485799-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR